missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HMGB1 / HMG1 Protein

A DNA sequence encoding the Homo sapiens (Human) HMGB1 / HMG1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9179-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067781

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001300821.1
HMGB1 / HMG1 Protein
Research Use Only
Hmgb1
Recombinant Protein
Yeast
GKGDPKKPRG KMSSYAFFVQ TCREEHKKKH PDASVNFSEF SKKCSERWKT MSAKEKGKFE DMAKADKARY EREMKTYIPP KGETKKKFKD PNAPKRPPSA FFLFCSEYRP KIKGEHPGLS IGDVAKKLGE MWNNTAADDK QPYEKKAAKL KEKYEKDIAA YRAKGKPDAA KKGVVKAEKS KKKKEEEEDE EDEEDEEEEE DEEDEDEEED DDDE
Greater than 90% as determined by SDS-PAGE.
3146
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers