missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGB1 / HMG1 Protein
A DNA sequence encoding the Homo sapiens (Human) HMGB1 / HMG1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9179-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001300821.1 | |
HMGB1 / HMG1 Protein | |
Research Use Only | |
Hmgb1 | |
Recombinant Protein | |
Yeast | |
GKGDPKKPRG KMSSYAFFVQ TCREEHKKKH PDASVNFSEF SKKCSERWKT MSAKEKGKFE DMAKADKARY EREMKTYIPP KGETKKKFKD PNAPKRPPSA FFLFCSEYRP KIKGEHPGLS IGDVAKKLGE MWNNTAADDK QPYEKKAAKL KEKYEKDIAA YRAKGKPDAA KKGVVKAEKS KKKKEEEEDE EDEEDEEEEE DEEDEDEEED DDDE | |
Greater than 90% as determined by SDS-PAGE. |
3146 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |