missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-B Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8824-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
HLA-B Protein | |
Research Use Only | |
HLA-B | |
Recombinant Protein | |
E. coli | |
GSHSMRYFYT AMSRPGRGEP RFISVGYVDD TQFVRFDSDA ASPREEPRAP WIEQEGPEYW DRNTQICKTN TQTYRESLRN LRGYYNQSEA GSHTLQRMYG CDVGPDGRLL RGHDQYAYDG KDYIALNEDL SSWTAADTAA QITQRKWEAA REAEQLRAYL EGLCVEWLRR YLENGKETLQ RADPPKTHVT HHPISDHEAT LRCWALGFYP AEITLTWQRD GEDQTQDTEL VETRPAGDRT FQKWAAVVVP SGEEQRYTCH VQHEGLPKPL TLRWEPSSQS TIPI | |
Greater than 80% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |