missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-B Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-B, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Name | HLA-B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HLA-B |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086671
|
enQuireBio™
QP8824-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006681
|
enQuireBio™
QP8824-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096671
|
enQuireBio™
QP8824-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016681
|
enQuireBio™
QP8824-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026681
|
enQuireBio™
QP8824-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036681
|
enQuireBio™
QP8824-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
HLA-B Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HLA-B | |
Recombinant Protein | |
E. coli | |
GSHSMRYFYT AMSRPGRGEP RFISVGYVDD TQFVRFDSDA ASPREEPRAP WIEQEGPEYW DRNTQICKTN TQTYRESLRN LRGYYNQSEA GSHTLQRMYG CDVGPDGRLL RGHDQYAYDG KDYIALNEDL SSWTAADTAA QITQRKWEAA REAEQLRAYL EGLCVEWLRR YLENGKETLQ RADPPKTHVT HHPISDHEAT LRCWALGFYP AEITLTWQRD GEDQTQDTEL VETRPAGDRT FQKWAAVVVP SGEEQRYTCH VQHEGLPKPL TLRWEPSSQS TIPI | |
Greater than 80% as determined by SDS-PAGE. |