missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein

A DNA sequence encoding the Homo sapiens (Human) HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7356-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028410

  • 2090.00 EUR / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057856.1
HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein
Research Use Only
gp120
Recombinant Protein
E. coli
KLWVTVYYGV PVWKEATTTL FCASDAKAYD TEVHNVWATH ACVPTDPNPQ EVVLVNVTEN FNMWKNDMVE QMHEDIISLW DQSLKPCVKL TPLCVSLKCT DLKNDTNTNS SSGRMIMEKG EIKNCSFNIS TSIRGKVQKE YAFFYKLDII PIDNDTTSYK LTSCNTSVIT QACPKVSFEP IPIHYCAPAG FAILKCNNKT FNGTGPCTNV STVQCTHGIR PVVSTQLLLN GSLAEEEVVI RSVNFTDNAK TIIVQLNTSV EINCTRPNNN TRKRIRIQRG PGRAFVTIGK IGNMRQAHCN ISRAKWNNTL KQIASKLREQ FGNNKTIIFK QSSGGDPEIV THSFNCGGEF FYCNSTQLFN STWFNSTWST EGSNNTEGSD TITLPCRIKQ IINMWQKVGK AMYAPPISGQ IRCSSNITGL LLTRDGGNSN NESEIFRPGG GDMRDNWRSE LYKYKVVKIE PLGVAPTKAK RRVVQREKR
Greater than 90% as determined by SDS-PAGE.
155971
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers