missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein
A DNA sequence encoding the Homo sapiens (Human) HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_057856.1 |
---|---|
Gene ID (Entrez) | 155971 |
Name | HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008410
|
enQuireBio™
QP7356-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028410
|
enQuireBio™
QP7356-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018410
|
enQuireBio™
QP7356-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038410
|
enQuireBio™
QP7356-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048410
|
enQuireBio™
QP7356-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058410
|
enQuireBio™
QP7356-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057856.1 | |
HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
155971 | |
Research Use Only | |
gp120 | |
Recombinant Protein | |
E. coli | |
KLWVTVYYGV PVWKEATTTL FCASDAKAYD TEVHNVWATH ACVPTDPNPQ EVVLVNVTEN FNMWKNDMVE QMHEDIISLW DQSLKPCVKL TPLCVSLKCT DLKNDTNTNS SSGRMIMEKG EIKNCSFNIS TSIRGKVQKE YAFFYKLDII PIDNDTTSYK LTSCNTSVIT QACPKVSFEP IPIHYCAPAG FAILKCNNKT FNGTGPCTNV STVQCTHGIR PVVSTQLLLN GSLAEEEVVI RSVNFTDNAK TIIVQLNTSV EINCTRPNNN TRKRIRIQRG PGRAFVTIGK IGNMRQAHCN ISRAKWNNTL KQIASKLREQ FGNNKTIIFK QSSGGDPEIV THSFNCGGEF FYCNSTQLFN STWFNSTWST EGSNNTEGSD TITLPCRIKQ IINMWQKVGK AMYAPPISGQ IRCSSNITGL LLTRDGGNSN NESEIFRPGG GDMRDNWRSE LYKYKVVKIE PLGVAPTKAK RRVVQREKR | |
Greater than 90% as determined by SDS-PAGE. |