missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone acetyltransferase KAT5 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone acetyltransferase KAT5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8905-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001193762.1 | |
Histone acetyltransferase KAT5 Protein | |
Research Use Only | |
KAT5 | |
Recombinant Protein | |
E. coli | |
EVGEIIEGCR LPVLRRNQDN EDEWPLAEIL SVKDISGRKL FYVHYIDFNK RLDEWVTHER LDLKKIQFPK KEAKTPTKNG LPGSRPGSPE REVPASAQAS GKTLPIPVQI TLRFNLPKER EAIPGGEPDQ PLSSSSCLQP NHRSTKRKVE VVSPATPVPS ETAPASVFPQ NGAARRAVAA QPGRKRKSNC LGTDEDSQDS SDGIPSAPRM TGSLVSDRSH DDIVTRMKNI ECIELGRHRL KPWYFSPYPQ ELTTLPVLYL CEFCLKYGRS LKCLQRHLTK CDLRHPPGNE IYRKGTISFF EIDGRKNKSY SQNLCLLAKC FLDHKTLYYD TDPFLFYVMT EYDCKGFHIV GYFSKEKEST EDYNVACILT LPPYQRRGYG KLLIEFSYEL SKVEGKTGTP EKPLSDLGLL SYRSYWSQTI LEILMGLKSE SGERPQITIN EISEITSIKK EDVISTLQYL NLINYYKGQY ILTLSEDIVD GHERAMLKRL LRIDSKCLHF TPKDWSKRGK W | |
Greater than 90% as determined by SDS-PAGE. |
10524 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |