missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone acetyltransferase KAT5 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone acetyltransferase KAT5, was expressed in the hosts and tags indicated.
186.00 EUR - 1855.00 EUR
Specifications
Accession Number | NP_001193762.1 |
---|---|
Gene ID (Entrez) | 10524 |
Name | Histone acetyltransferase KAT5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047111
|
enQuireBio™
QP8905-EC-10UG |
10 μg |
186.00 EUR
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067111
|
enQuireBio™
QP8905-EC-200UG |
200 μg |
765.00 EUR
200µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037111
|
enQuireBio™
QP8905-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087111
|
enQuireBio™
QP8905-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077111
|
enQuireBio™
QP8905-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057111
|
enQuireBio™
QP8905-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001193762.1 | |
Histone acetyltransferase KAT5 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10524 | |
Research Use Only | |
KAT5 | |
Recombinant Protein | |
E. coli | |
EVGEIIEGCR LPVLRRNQDN EDEWPLAEIL SVKDISGRKL FYVHYIDFNK RLDEWVTHER LDLKKIQFPK KEAKTPTKNG LPGSRPGSPE REVPASAQAS GKTLPIPVQI TLRFNLPKER EAIPGGEPDQ PLSSSSCLQP NHRSTKRKVE VVSPATPVPS ETAPASVFPQ NGAARRAVAA QPGRKRKSNC LGTDEDSQDS SDGIPSAPRM TGSLVSDRSH DDIVTRMKNI ECIELGRHRL KPWYFSPYPQ ELTTLPVLYL CEFCLKYGRS LKCLQRHLTK CDLRHPPGNE IYRKGTISFF EIDGRKNKSY SQNLCLLAKC FLDHKTLYYD TDPFLFYVMT EYDCKGFHIV GYFSKEKEST EDYNVACILT LPPYQRRGYG KLLIEFSYEL SKVEGKTGTP EKPLSDLGLL SYRSYWSQTI LEILMGLKSE SGERPQITIN EISEITSIKK EDVISTLQYL NLINYYKGQY ILTLSEDIVD GHERAMLKRL LRIDSKCLHF TPKDWSKRGK W | |
Greater than 90% as determined by SDS-PAGE. |