Learn More
R&D Systems™ Recombinant Human His6-alpha-Synuclein Protein
Click to view available options
:
500ug; Carrier Free
Descrizione
The Recombinant Human His6-Synuclein-alpha Protein is derived from E. coli. The Recombinant Human His6-Synuclein-alpha Protein has been validated for the following applications: Bioactivity.
Specifica
Specifica
| Accession Number | P37840.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES and NaCl. |
| Gene ID (Entrez) | 6622 |
| Molecular Weight (g/mol) | 15 kDa |
| Name | His6 Synuclein-alpha |
| Quantity | 500μg |
| Source | E. coli-derived human alpha-Synuclein protein Met1 - Ala140 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto