missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GPT2 His Protein

Product Code. p-7249164
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
20µg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18361246

Brand: Novus Biologicals™ NBP31818250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specific activity is > 100units/mg, and is defined as the amount of enzyme that cleaves 1 umole of L-Alanine to L-Glutamate per minute at pH 7.5 at 37C
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 30% glycerol, 2mM DTT, 0.2M NaCl
Gene ID (Entrez) 84706
Molecular Weight (g/mol) M.W. Theoretical: 60.3 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias AAT2, alanine aminotransferase 2, ALT2EC 2.6.1.2, Glutamate pyruvate transaminase 2, glutamic pyruvate transaminase (alanine aminotransferase) 2, Glutamic--alanine transaminase 2, glutamic-pyruvate transaminase 2, Glutamic--pyruvic transaminase 2, GPT 2
Gene Symbol GPT2
Research Category metabolism
Purity or Quality Grade >90%, by SDS-PAGE
Protein GPT2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.