missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Glucosylceramidase/GBA Protein
Click to view available options
Quantity:
1 mg
20 μg
Unit Size:
1mg
20µg
Description
Measured by its ability to hydrolyze 4-methylumbelliferyl-beta-D-glucopyranoside. The specific activity is >200 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P04062.3 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Citrate. |
| Gene ID (Entrez) | 2629 |
| Molecular Weight (g/mol) | M.W. (Observed): 60-75 kDa, reducing conditions; M.W. (theoretical): 56 kDa |
| Name | Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta; acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Glucosylceramidase/GBA protein Ala40-Gln536, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction