missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Glucosylceramidase/GBA Protein

Product Code. 18783969
Click to view available options
Quantity:
1 mg
20 μg
Unit Size:
1mg
20µg
This item is not returnable. View return policy

Product Code. 18783969

Brand: R&D Systems™ 7410GHB01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to hydrolyze 4-methylumbelliferyl-beta-D-glucopyranoside. The specific activity is >200 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04062.3
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Sodium Citrate.
Gene ID (Entrez) 2629
Molecular Weight (g/mol) M.W. (Observed): 60-75 kDa, reducing conditions; M.W. (theoretical): 56 kDa
Name Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta; acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived human Glucosylceramidase/GBA protein Ala40-Gln536, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Glucosylceramidase/GBA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.