missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Genome polyProtein

A DNA sequence encoding the Homo sapiens (Human) Genome polyprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7370-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028510

  • 463.00 EUR / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Genome polyProtein
Research Use Only
VP1
Recombinant Protein
E. coli
NPVENYIDSV LNEVLVVPNI QPSTSVSSHA APALDAAETG HTSSVQPEDM IETRYVITDQ TRDETSIESF LGRSGCIAMI EFNTSSDKTE HDKIGKGFKT WKVSLQEMAQ IRRKYELFTY TRFDSEITIV TAAAAQGNDS GHIVLQFMYV PPGAPVPEKR DDYTWQSGTN ASVFWQEGQP YPRFTIPFMS IASAYYMFYD GYDGDSAASK YGSVVTNDMG TICVRIVTSN QKHDSNIVCR IYHKAKHIKA WCPRPPRAVA YQHTHSTNYI PSNGEATTQI KTRPDVFTVT NV
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers