missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Genome polyProtein

A DNA sequence encoding the Homo sapiens (Human) Genome polyprotein, was expressed in the hosts and tags indicated.

160.00 EUR - 2090.00 EUR

Specifications

Name Genome polyProtein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol VP1
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16028510
View Documents Promotion Details
enQuireBio™
QP7370-EC-100UG
100 μg
463.00 EUR
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16048510
View Documents Promotion Details
enQuireBio™
QP7370-EC-1MG
1 mg
2090.00 EUR
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16038510
View Documents Promotion Details
enQuireBio™
QP7370-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16058510
View Documents Promotion Details
enQuireBio™
QP7370-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16068510
View Documents Promotion Details
enQuireBio™
QP7370-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16078510
View Documents Promotion Details
enQuireBio™
QP7370-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Genome polyProtein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Research Use Only
VP1
Recombinant Protein
E. coli
NPVENYIDSV LNEVLVVPNI QPSTSVSSHA APALDAAETG HTSSVQPEDM IETRYVITDQ TRDETSIESF LGRSGCIAMI EFNTSSDKTE HDKIGKGFKT WKVSLQEMAQ IRRKYELFTY TRFDSEITIV TAAAAQGNDS GHIVLQFMYV PPGAPVPEKR DDYTWQSGTN ASVFWQEGQP YPRFTIPFMS IASAYYMFYD GYDGDSAASK YGSVVTNDMG TICVRIVTSN QKHDSNIVCR IYHKAKHIKA WCPRPPRAVA YQHTHSTNYI PSNGEATTQI KTRPDVFTVT NV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications