missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galectin-9 Protein

A DNA sequence encoding the Homo sapiens (Human) Galectin-9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6300-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022290

  • 463.00 EUR / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001317092.1
Galectin-9 Protein
Research Use Only
LGALS9
Recombinant Protein
E. coli
MAFSGSQAPY LSPAVPFSGT IQGGLQDGLQ ITVNGTVLSS SGTRFAVNFQ TGFSGNDIAF HFNPRFEDGG YVVCNTRQNG SWGPEERKTH MPFQKGMPFD LCFLVQSSDF KVMVNGILFV QYFHRVPFHR VDTISVNGSV QLSYISFQPP GVWPANPAPI TQTVIHTVQS APGQMFSTPA IPPMMYPHPA YPMPFITTIL GGLYPSKSIL LSGTVLPSAQ RFHINLCSGN HIAFHLNPRF DENAVVRNTQ IDNSWGSEER SLPRKMPFVR GQSFSVWILC EAHCLKVAVD GQHLFEYYHR LRNLPTINRL EVGGDIQLTH VQT
Greater than 90% as determined by SDS-PAGE.
3965
100 μg
Not determined.
Full Length of Isoform 2
Human
His-GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers