missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galectin-9 Protein
A DNA sequence encoding the Homo sapiens (Human) Galectin-9, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001317092.1 |
---|---|
Gene ID (Entrez) | 3965 |
Name | Galectin-9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022290
|
enQuireBio™
QP6300-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042290
|
enQuireBio™
QP6300-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032290
|
enQuireBio™
QP6300-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052290
|
enQuireBio™
QP6300-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062290
|
enQuireBio™
QP6300-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072290
|
enQuireBio™
QP6300-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001317092.1 | |
Galectin-9 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-GST | |
Tris-based buffer, 50% glycerol |
3965 | |
Research Use Only | |
LGALS9 | |
Recombinant Protein | |
E. coli | |
MAFSGSQAPY LSPAVPFSGT IQGGLQDGLQ ITVNGTVLSS SGTRFAVNFQ TGFSGNDIAF HFNPRFEDGG YVVCNTRQNG SWGPEERKTH MPFQKGMPFD LCFLVQSSDF KVMVNGILFV QYFHRVPFHR VDTISVNGSV QLSYISFQPP GVWPANPAPI TQTVIHTVQS APGQMFSTPA IPPMMYPHPA YPMPFITTIL GGLYPSKSIL LSGTVLPSAQ RFHINLCSGN HIAFHLNPRF DENAVVRNTQ IDNSWGSEER SLPRKMPFVR GQSFSVWILC EAHCLKVAVD GQHLFEYYHR LRNLPTINRL EVGGDIQLTH VQT | |
Greater than 90% as determined by SDS-PAGE. |