missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G-CSFR / CD114 Protein
A DNA sequence encoding the Homo sapiens (Human) G-CSFR / CD114, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8705-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000751.1 | |
G-CSFR / CD114 Protein | |
Research Use Only | |
CSF3R | |
Recombinant Protein | |
E. coli | |
ECGHISVSAP IVHLGDPITA SCIIKQNCSH LDPEPQILWR LGAELQPGGR QQRLSDGTQE SIITLPHLNH TQAFLSCCLN WGNSLQILDQ VELRAGYPPA IPHNLSCLMN LTTSSLICQW EPGPETHLPT SFTLKSFKSR GNCQTQGDSI LDCVPKDGQS HCCIPRKHLL LYQNMGIWVQ AENALGTSMS PQLCLDPMDV VKLEPPMLRT MDPSPEAAPP QAGCLQLCWE PWQPGLHINQ KCELRHKPQR GEASWALVGP LPLEALQYEL CGLLPATAYT LQIRCIRWPL PGHWSDWSPS LELRTTERAP TVRLDTWWRQ RQLDPRTVQL FWKPVPLEED SGRIQGYVVS WRPSGQAGAI LPLCNTTELS CTFHLPSEAQ EVALVAYNSA GTSRPTPVVF SESRGPALTR LHAMARDPHS LWVGWEPPNP WPQGYVIEWG LGPPSASNSN KTWRMEQNGR ATGFLLKENI RPFQLYEIIV TPLYQDTMGP SQHVYAYSQE MAPSHAPELH LKHIGKTWAQ LEWVPEPPEL GKSPLTHYTI FWTNAQNQSF SAILNASSRG FVLHGLEPAS LYHIHLMAAS QAGATNSTVL TLMTLT | |
Greater than 90% as determined by SDS-PAGE. |
1441 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |