missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G-CSFR / CD114 Protein
A DNA sequence encoding the Homo sapiens (Human) G-CSFR / CD114, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_000751.1 |
---|---|
Gene ID (Entrez) | 1441 |
Name | G-CSFR / CD114 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086041
|
enQuireBio™
QP8705-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006051
|
enQuireBio™
QP8705-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096041
|
enQuireBio™
QP8705-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016051
|
enQuireBio™
QP8705-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026051
|
enQuireBio™
QP8705-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036051
|
enQuireBio™
QP8705-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000751.1 | |
G-CSFR / CD114 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1441 | |
Research Use Only | |
CSF3R | |
Recombinant Protein | |
E. coli | |
ECGHISVSAP IVHLGDPITA SCIIKQNCSH LDPEPQILWR LGAELQPGGR QQRLSDGTQE SIITLPHLNH TQAFLSCCLN WGNSLQILDQ VELRAGYPPA IPHNLSCLMN LTTSSLICQW EPGPETHLPT SFTLKSFKSR GNCQTQGDSI LDCVPKDGQS HCCIPRKHLL LYQNMGIWVQ AENALGTSMS PQLCLDPMDV VKLEPPMLRT MDPSPEAAPP QAGCLQLCWE PWQPGLHINQ KCELRHKPQR GEASWALVGP LPLEALQYEL CGLLPATAYT LQIRCIRWPL PGHWSDWSPS LELRTTERAP TVRLDTWWRQ RQLDPRTVQL FWKPVPLEED SGRIQGYVVS WRPSGQAGAI LPLCNTTELS CTFHLPSEAQ EVALVAYNSA GTSRPTPVVF SESRGPALTR LHAMARDPHS LWVGWEPPNP WPQGYVIEWG LGPPSASNSN KTWRMEQNGR ATGFLLKENI RPFQLYEIIV TPLYQDTMGP SQHVYAYSQE MAPSHAPELH LKHIGKTWAQ LEWVPEPPEL GKSPLTHYTI FWTNAQNQSF SAILNASSRG FVLHGLEPAS LYHIHLMAAS QAGATNSTVL TLMTLT | |
Greater than 90% as determined by SDS-PAGE. |