missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human G-CSF Protein

Product Code. 18117570
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Acetic Acid.
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 18117570

Brand: R&D Systems™ 214CS025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human G-CSF Protein is derived from E. coli. The Recombinant Human G-CSF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in Acetic Acid.
Gene ID (Entrez) 1440
Molecular Weight (g/mol) 18.8 kDa
Name G-CSF
Quantity 25 μg
Source E. coli-derived human G-CSF protein Thr31-Pro204,with an N-terminal Met
Storage Requirements Do not freeze. 6 months from date of receipt, 2 to 8° C as supplied.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.