missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOXP3 / Scurfin Protein
A DNA sequence encoding the Homo sapiens (Human) FOXP3 / Scurfin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8828-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001107849.1 | |
FOXP3 / Scurfin Protein | |
Research Use Only | |
FOXP3 | |
Recombinant Protein | |
E. coli | |
MPNPRPGKPS APSLALGPSP GASPSWRAAP KASDLLGARG PGGTFQGRDL RGGAHASSSS LNPMPPSQLQ LPTLPLVMVA PSGARLGPLP HLQALLQDRP HFMHQLSTVD AHARTPVLQV HPLESPAMIS LTPPTTATGV FSLKARPGLP PGINVASLEW VSREPALLCT FPNPSAPRKD STLSAVPQSS YPLLANGVCK WPGCEKVFEE PEDFLKHCQA DHLLDEKGRA QCLLQREMVQ SLEQQLVLEK EKLSAMQAHL | |
Greater than 90% as determined by SDS-PAGE. |
50943 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |