missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOXP3 / Scurfin Protein
A DNA sequence encoding the Homo sapiens (Human) FOXP3 / Scurfin, was expressed in the hosts and tags indicated.
160.00 EUR - 1855.00 EUR
Specifications
Accession Number | NP_001107849.1 |
---|---|
Gene ID (Entrez) | 50943 |
Name | FOXP3 / Scurfin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16066691
|
enQuireBio™
QP8828-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006701
|
enQuireBio™
QP8828-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056691
|
enQuireBio™
QP8828-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086691
|
enQuireBio™
QP8828-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096691
|
enQuireBio™
QP8828-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076691
|
enQuireBio™
QP8828-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001107849.1 | |
FOXP3 / Scurfin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
50943 | |
Research Use Only | |
FOXP3 | |
Recombinant Protein | |
E. coli | |
MPNPRPGKPS APSLALGPSP GASPSWRAAP KASDLLGARG PGGTFQGRDL RGGAHASSSS LNPMPPSQLQ LPTLPLVMVA PSGARLGPLP HLQALLQDRP HFMHQLSTVD AHARTPVLQV HPLESPAMIS LTPPTTATGV FSLKARPGLP PGINVASLEW VSREPALLCT FPNPSAPRKD STLSAVPQSS YPLLANGVCK WPGCEKVFEE PEDFLKHCQA DHLLDEKGRA QCLLQREMVQ SLEQQLVLEK EKLSAMQAHL | |
Greater than 90% as determined by SDS-PAGE. |