Learn More
enQuireBio™ Recombinant Human FLT3L / Flt3 ligand Protein
Product Code. 15905919
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15905919 500 μg 500µg
15995909 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15905919 Supplier enQuireBio™ Supplier No. QP5327500ug
Recombinant Protein
Specifications
Accession Number NP_001191431.1
Gene ID (Entrez) 2323
Name FLT3L / Flt3 ligand Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol FLT3LG
Biological Activity Shows bioactivity at a concentration of 10 ng/mL in the OCI-AML5 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MTQDCSFQHS PISSDFAVKI RELSDYLLQD YPVTVASNLQ DEELCGGLWR LVLAQRWMER LKTVAGSKMQ GLLERVNTEI HFVTKCAFQP PPSCLRFVQT NISRLLQETS EQLVALKPWI TRQNFSRCLE LQCQPDSSTL PPPWSPRPLE ATAPT
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.