missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Estradiol 17-beta-dehydrogenase 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Estradiol 17-beta-dehydrogenase 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7921-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057329.3 | |
Estradiol 17-beta-dehydrogenase 11 Protein | |
Research Use Only | |
HSD17B11 | |
Recombinant Protein | |
E. coli | |
ESFVKLFIPK RRKSVTGEIV LITGAGHGIG RLTAYEFAKL KSKLVLWDIN KHGLEETAAK CKGLGAKVHT FVVDCSNRED IYSSAKKVKA EIGDVSILVN NAGVVYTSDL FATQDPQIEK TFEVNVLAHF WTTKAFLPAM TKNNHGHIVT VASAAGHVSV PFLLAYCSSK FAAVGFHKTL TDELAALQIT GVKTTCLCPN FVNTGFIKNP STSLGPTLEP EEVVNRLMHG ILTEQKMIFI PSSIAFLTTL ERILPERFLA VLKQKISVKF DAVIGYKMKA Q | |
Greater than 90% as determined by SDS-PAGE. |
51170 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |