missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Estradiol 17-beta-dehydrogenase 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Estradiol 17-beta-dehydrogenase 11, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_057329.3 |
---|---|
Gene ID (Entrez) | 51170 |
Name | Estradiol 17-beta-dehydrogenase 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011601
|
enQuireBio™
QP7921-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031601
|
enQuireBio™
QP7921-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021601
|
enQuireBio™
QP7921-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041601
|
enQuireBio™
QP7921-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051601
|
enQuireBio™
QP7921-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061601
|
enQuireBio™
QP7921-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057329.3 | |
Estradiol 17-beta-dehydrogenase 11 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
51170 | |
Research Use Only | |
HSD17B11 | |
Recombinant Protein | |
E. coli | |
ESFVKLFIPK RRKSVTGEIV LITGAGHGIG RLTAYEFAKL KSKLVLWDIN KHGLEETAAK CKGLGAKVHT FVVDCSNRED IYSSAKKVKA EIGDVSILVN NAGVVYTSDL FATQDPQIEK TFEVNVLAHF WTTKAFLPAM TKNNHGHIVT VASAAGHVSV PFLLAYCSSK FAAVGFHKTL TDELAALQIT GVKTTCLCPN FVNTGFIKNP STSLGPTLEP EEVVNRLMHG ILTEQKMIFI PSSIAFLTTL ERILPERFLA VLKQKISVKF DAVIGYKMKA Q | |
Greater than 90% as determined by SDS-PAGE. |