missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Embryonic stem cell-related gene Protein

A DNA sequence encoding the Homo sapiens (Human) Embryonic stem cell-related gene protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7626-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049870

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Embryonic stem cell-related gene Protein
Research Use Only
ESRG
Recombinant Protein
E. coli
MTLFSDSARL HPGEINSLVA HTKPVWWSLH TDAHEIWCRD SDRGTSLGRS IPCPPALCSV RKIHLRPQVL RPTSPRNISP ISNPVSGLFL LCSPTSLTIP QPLSPFNLGA TLQSLPSLNF NSFHSLVETK ETCFIREPKT PAPVTDWEGS LPLVFNHCRD ASLISRFRPR RDACLGPSPL AASPAFLGQG QVPLNPFSFT LSGKSRFSGA GASTPQPLLL HP
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers