missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Embryonic stem cell-related gene Protein
A DNA sequence encoding the Homo sapiens (Human) Embryonic stem cell-related gene protein, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Name | Embryonic stem cell-related gene Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | ESRG |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019870
|
enQuireBio™
QP7626-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039870
|
enQuireBio™
QP7626-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16029870
|
enQuireBio™
QP7626-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049870
|
enQuireBio™
QP7626-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059870
|
enQuireBio™
QP7626-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069870
|
enQuireBio™
QP7626-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Embryonic stem cell-related gene Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
ESRG | |
Recombinant Protein | |
E. coli | |
MTLFSDSARL HPGEINSLVA HTKPVWWSLH TDAHEIWCRD SDRGTSLGRS IPCPPALCSV RKIHLRPQVL RPTSPRNISP ISNPVSGLFL LCSPTSLTIP QPLSPFNLGA TLQSLPSLNF NSFHSLVETK ETCFIREPKT PAPVTDWEGS LPLVFNHCRD ASLISRFRPR RDACLGPSPL AASPAFLGQG QVPLNPFSFT LSGKSRFSGA GASTPQPLLL HP | |
Greater than 90% as determined by SDS-PAGE. |