missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF3F Protein

A DNA sequence encoding the Homo sapiens (Human) EIF3F, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8436-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024611

  • 2090.00 EUR / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003745.1
EIF3F Protein
Research Use Only
EIF3F
Recombinant Protein
E. coli
MATPAVPVSA PPATPTPVPA AAPASVPAPT PAPAAAPVPA AAPASSSDPA AAAAATAAPG QTPASAQAPA QTPAPALPGP ALPGPFPGGR VVRLHPVILA SIVDSYERRN EGAARVIGTL LGTVDKHSVE VTNCFSVPHN ESEDEVAVDM EFAKNMYELH KKVSPNELIL GWYATGHDIT EHSVLIHEYY SREAPNPIHL TVDTSLQNGR MSIKAYVSTL MGVPGRTMGV MFTPLTVKYA YYDTERIGVD LIMKTCFSPN RVIGLSSDLQ QVGGASARIQ DALSTVLQYA EDVLSGKVSA DNTVGRFLMS LVNQVPKIVP DDF
Greater than 90% as determined by SDS-PAGE.
8665
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers