missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3F Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3F, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_003745.1 |
---|---|
Gene ID (Entrez) | 8665 |
Name | EIF3F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004611
|
enQuireBio™
QP8436-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024611
|
enQuireBio™
QP8436-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014611
|
enQuireBio™
QP8436-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034611
|
enQuireBio™
QP8436-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044611
|
enQuireBio™
QP8436-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054611
|
enQuireBio™
QP8436-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003745.1 | |
EIF3F Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
8665 | |
Research Use Only | |
EIF3F | |
Recombinant Protein | |
E. coli | |
MATPAVPVSA PPATPTPVPA AAPASVPAPT PAPAAAPVPA AAPASSSDPA AAAAATAAPG QTPASAQAPA QTPAPALPGP ALPGPFPGGR VVRLHPVILA SIVDSYERRN EGAARVIGTL LGTVDKHSVE VTNCFSVPHN ESEDEVAVDM EFAKNMYELH KKVSPNELIL GWYATGHDIT EHSVLIHEYY SREAPNPIHL TVDTSLQNGR MSIKAYVSTL MGVPGRTMGV MFTPLTVKYA YYDTERIGVD LIMKTCFSPN RVIGLSSDLQ QVGGASARIQ DALSTVLQYA EDVLSGKVSA DNTVGRFLMS LVNQVPKIVP DDF | |
Greater than 90% as determined by SDS-PAGE. |