missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3F Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3F, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8436-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003745.1 | |
EIF3F Protein | |
Research Use Only | |
EIF3F | |
Recombinant Protein | |
E. coli | |
MATPAVPVSA PPATPTPVPA AAPASVPAPT PAPAAAPVPA AAPASSSDPA AAAAATAAPG QTPASAQAPA QTPAPALPGP ALPGPFPGGR VVRLHPVILA SIVDSYERRN EGAARVIGTL LGTVDKHSVE VTNCFSVPHN ESEDEVAVDM EFAKNMYELH KKVSPNELIL GWYATGHDIT EHSVLIHEYY SREAPNPIHL TVDTSLQNGR MSIKAYVSTL MGVPGRTMGV MFTPLTVKYA YYDTERIGVD LIMKTCFSPN RVIGLSSDLQ QVGGASARIQ DALSTVLQYA EDVLSGKVSA DNTVGRFLMS LVNQVPKIVP DDF | |
Greater than 90% as determined by SDS-PAGE. |
8665 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |