missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF / Epidermal Growth Factor Protein
A DNA sequence encoding the Human EGF / Epidermal Growth Factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10394-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171601.1 | |
EGF / Epidermal Growth Factor Protein | |
Research Use Only | |
EGF | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 m filtered 20 mM PBS, pH 7.2, lyophilized |
1950 | |
500 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR | |
Greater than 98% as determined by SDS-PAGE and HPLC. |