missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF / Epidermal Growth Factor Protein
A DNA sequence encoding the Human EGF / Epidermal Growth Factor, was expressed in the hosts and tags indicated.
116.00 EUR - 299.00 EUR
Specifications
Accession Number | NP_001171601.1 |
---|---|
Gene ID (Entrez) | 1950 |
Name | EGF / Epidermal Growth Factor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15903718
|
enQuireBio™
QP10394-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15913718
|
enQuireBio™
QP10394-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001171601.1 | |
EGF / Epidermal Growth Factor Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR | |
Greater than 98% as determined by SDS-PAGE and HPLC. |
1950 | |
Research Use Only | |
EGF | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 m filtered 20 mM PBS, pH 7.2, lyophilized |