missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase MIB1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase MIB1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_065825.1 |
---|---|
Gene ID (Entrez) | 57534 |
Name | E3 ubiquitin-protein ligase MIB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16066331
|
enQuireBio™
QP8760-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086331
|
enQuireBio™
QP8760-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076331
|
enQuireBio™
QP8760-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096331
|
enQuireBio™
QP8760-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006341
|
enQuireBio™
QP8760-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016341
|
enQuireBio™
QP8760-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_065825.1 | |
E3 ubiquitin-protein ligase MIB1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
57534 | |
Research Use Only | |
MIB1 | |
Recombinant Protein | |
E. coli | |
RNNRVMVEGV GARVVRGPDW KWGKQDGGEG HVGTVRSFES PEEVVVVWDN GTAANYRCSG AYDLRILDSA PTGIKHDGTM CDTCRQQPII GIRWKCAECT NYDLCTVCYH GDKHHLRHRF YRITTPGSER VLLESRRKSK KITARGIFAG ARVVRGVDWQ WEDQDGGNGR RGKVTEIQDW SASSPHSAAY VLWDNGAKNL YRVGFEGMSD LKCVQDAKGG SFYRDHCPVL GEQNGNRNPG GLQIGDLVNI DLDLEIVQSL QHGHGGWTDG MFETLTTTGT VCGIDEDHDI VVQYPSGNRW TFNPAVLTKA NIVRSGDAAQ GAEGGTSQ | |
Greater than 90% as determined by SDS-PAGE. |