missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase MIB1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase MIB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8760-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065825.1 | |
E3 ubiquitin-protein ligase MIB1 Protein | |
Research Use Only | |
MIB1 | |
Recombinant Protein | |
E. coli | |
RNNRVMVEGV GARVVRGPDW KWGKQDGGEG HVGTVRSFES PEEVVVVWDN GTAANYRCSG AYDLRILDSA PTGIKHDGTM CDTCRQQPII GIRWKCAECT NYDLCTVCYH GDKHHLRHRF YRITTPGSER VLLESRRKSK KITARGIFAG ARVVRGVDWQ WEDQDGGNGR RGKVTEIQDW SASSPHSAAY VLWDNGAKNL YRVGFEGMSD LKCVQDAKGG SFYRDHCPVL GEQNGNRNPG GLQIGDLVNI DLDLEIVQSL QHGHGGWTDG MFETLTTTGT VCGIDEDHDI VVQYPSGNRW TFNPAVLTKA NIVRSGDAAQ GAEGGTSQ | |
Greater than 90% as determined by SDS-PAGE. |
57534 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |