missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA repair protein complementing XP-C cells Protein

A DNA sequence encoding the Homo sapiens (Human) DNA repair protein complementing XP-C cells, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6892-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026600

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_004619.3
DNA repair protein complementing XP-C cells Protein
Research Use Only
XPC
Recombinant Protein
E. coli
SLPAASSSSS SSKRGKKMCS DGEKAEKRSI AGIDQWLEVF CEQEEKWVCV DCVHGVVGQP LTCYKYATKP MTYVVGIDSD GWVRDVTQRY DPVWMTVTRK CRVDAEWWAE TLRPYQSPFM DREKKEDLEF QAKHMDQPLP TAIGLYKNHP LYALKRHLLK YEAIYPETAA ILGYCRGEAV YSRDCVHTLH SRDTWLKKAR VVRLGEVPYK MVKGFSNRAR KARLAEPQLR EENDLGLFG
Greater than 90% as determined by SDS-PAGE.
7508
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers