missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein complementing XP-C cells Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein complementing XP-C cells, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_004619.3 |
---|---|
Gene ID (Entrez) | 7508 |
Name | DNA repair protein complementing XP-C cells Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096590
|
enQuireBio™
QP6892-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016600
|
enQuireBio™
QP6892-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006600
|
enQuireBio™
QP6892-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026600
|
enQuireBio™
QP6892-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036600
|
enQuireBio™
QP6892-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046600
|
enQuireBio™
QP6892-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004619.3 | |
DNA repair protein complementing XP-C cells Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7508 | |
Research Use Only | |
XPC | |
Recombinant Protein | |
E. coli | |
SLPAASSSSS SSKRGKKMCS DGEKAEKRSI AGIDQWLEVF CEQEEKWVCV DCVHGVVGQP LTCYKYATKP MTYVVGIDSD GWVRDVTQRY DPVWMTVTRK CRVDAEWWAE TLRPYQSPFM DREKKEDLEF QAKHMDQPLP TAIGLYKNHP LYALKRHLLK YEAIYPETAA ILGYCRGEAV YSRDCVHTLH SRDTWLKKAR VVRLGEVPYK MVKGFSNRAR KARLAEPQLR EENDLGLFG | |
Greater than 90% as determined by SDS-PAGE. |