missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytoplasmic protein NCK2 Protein

A DNA sequence encoding the Homo sapiens (Human) Cytoplasmic protein NCK2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6408-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16053100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001004720.1
Cytoplasmic protein NCK2 Protein
Research Use Only
NCK2
Recombinant Protein
E. coli
MTEEVIVIAK WDYTAQQDQE LDIKKNERLW LLDDSKTWWR VRNAANRTGY VPSNYVERKN SLKKGSLVKN LKDTLGLGKT RRKTSARDAS PTPSTDAEYP ANGSGADRIY DLNIPAFVKF AYVAEREDEL SLVKGSRVTV MEKCSDGWWR GSYNGQIGWF PSNYVLEEVD EAAAESPSFL SLRKGASLSN GQGSRVLHVV QTLYPFSSVT EEELNFEKGE TMEVIEKPEN DPEWWKCKNA RGQVGLVPKN YVVVLSDGPA LHPAHAPQIS YTGPSSSGRF AGREWYYGNV TRHQAECALN ERGVEGDFLI RDSESSPSDF SVSLKASGKN KHFKVQLVDN VYCIGQRRFH TMDELVEHYK KAPIFTSEHG EKLYLVRALQ
Greater than 90% as determined by SDS-PAGE.
8440
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers