missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytoplasmic protein NCK2 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytoplasmic protein NCK2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001004720.1 |
---|---|
Gene ID (Entrez) | 8440 |
Name | Cytoplasmic protein NCK2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023100
|
enQuireBio™
QP6408-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043100
|
enQuireBio™
QP6408-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033100
|
enQuireBio™
QP6408-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053100
|
enQuireBio™
QP6408-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063100
|
enQuireBio™
QP6408-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073100
|
enQuireBio™
QP6408-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001004720.1 | |
Cytoplasmic protein NCK2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8440 | |
Research Use Only | |
NCK2 | |
Recombinant Protein | |
E. coli | |
MTEEVIVIAK WDYTAQQDQE LDIKKNERLW LLDDSKTWWR VRNAANRTGY VPSNYVERKN SLKKGSLVKN LKDTLGLGKT RRKTSARDAS PTPSTDAEYP ANGSGADRIY DLNIPAFVKF AYVAEREDEL SLVKGSRVTV MEKCSDGWWR GSYNGQIGWF PSNYVLEEVD EAAAESPSFL SLRKGASLSN GQGSRVLHVV QTLYPFSSVT EEELNFEKGE TMEVIEKPEN DPEWWKCKNA RGQVGLVPKN YVVVLSDGPA LHPAHAPQIS YTGPSSSGRF AGREWYYGNV TRHQAECALN ERGVEGDFLI RDSESSPSDF SVSLKASGKN KHFKVQLVDN VYCIGQRRFH TMDELVEHYK KAPIFTSEHG EKLYLVRALQ | |
Greater than 90% as determined by SDS-PAGE. |