missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytoplasmic protein NCK2 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytoplasmic protein NCK2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6408-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001004720.1 | |
Cytoplasmic protein NCK2 Protein | |
Research Use Only | |
NCK2 | |
Recombinant Protein | |
E. coli | |
MTEEVIVIAK WDYTAQQDQE LDIKKNERLW LLDDSKTWWR VRNAANRTGY VPSNYVERKN SLKKGSLVKN LKDTLGLGKT RRKTSARDAS PTPSTDAEYP ANGSGADRIY DLNIPAFVKF AYVAEREDEL SLVKGSRVTV MEKCSDGWWR GSYNGQIGWF PSNYVLEEVD EAAAESPSFL SLRKGASLSN GQGSRVLHVV QTLYPFSSVT EEELNFEKGE TMEVIEKPEN DPEWWKCKNA RGQVGLVPKN YVVVLSDGPA LHPAHAPQIS YTGPSSSGRF AGREWYYGNV TRHQAECALN ERGVEGDFLI RDSESSPSDF SVSLKASGKN KHFKVQLVDN VYCIGQRRFH TMDELVEHYK KAPIFTSEHG EKLYLVRALQ | |
Greater than 90% as determined by SDS-PAGE. |
8440 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |