missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytoplasmic envelopment protein 1
A DNA sequence encoding the Homo sapiens (Human) Cytoplasmic envelopment protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8931-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Cytoplasmic envelopment protein 1 | |
Research Use Only | |
UL99 | |
Recombinant Protein | |
E. coli | |
GAELCKRICC EFGTTPGEPL KDALGRQVSL RSYDNIPPTS SSDEGEDDDD GEDDDNEERQ QKLRLCGSGC GGNDSSSGSH REATHDGSKK NAVRSTFRED KAPKPSKQSK KKKKPSKHHH HQQSSIMQET DDLDEEDTSI YLSPPPVPPV QVVAKRLPRP DTPRTPRQKK ISQRPPTPGT KKPAASLPF | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |