missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytoplasmic envelopment protein 1
A DNA sequence encoding the Homo sapiens (Human) Cytoplasmic envelopment protein 3, was expressed in the hosts and tags indicated.
160.00 EUR - 1855.00 EUR
Specifications
Name | Cytoplasmic envelopment protein 1 |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | UL99 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067211
|
enQuireBio™
QP8931-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007221
|
enQuireBio™
QP8931-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057211
|
enQuireBio™
QP8931-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087211
|
enQuireBio™
QP8931-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097211
|
enQuireBio™
QP8931-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077211
|
enQuireBio™
QP8931-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Cytoplasmic envelopment protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
UL99 | |
Recombinant Protein | |
E. coli | |
GAELCKRICC EFGTTPGEPL KDALGRQVSL RSYDNIPPTS SSDEGEDDDD GEDDDNEERQ QKLRLCGSGC GGNDSSSGSH REATHDGSKK NAVRSTFRED KAPKPSKQSK KKKKPSKHHH HQQSSIMQET DDLDEEDTSI YLSPPPVPPV QVVAKRLPRP DTPRTPRQKK ISQRPPTPGT KKPAASLPF | |
Greater than 90% as determined by SDS-PAGE. |