missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytoplasmic envelopment protein 1

A DNA sequence encoding the Homo sapiens (Human) Cytoplasmic envelopment protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8931-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077211

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Cytoplasmic envelopment protein 1
Research Use Only
UL99
Recombinant Protein
E. coli
GAELCKRICC EFGTTPGEPL KDALGRQVSL RSYDNIPPTS SSDEGEDDDD GEDDDNEERQ QKLRLCGSGC GGNDSSSGSH REATHDGSKK NAVRSTFRED KAPKPSKQSK KKKKPSKHHH HQQSSIMQET DDLDEEDTSI YLSPPPVPPV QVVAKRLPRP DTPRTPRQKK ISQRPPTPGT KKPAASLPF
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers