missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Collagen alpha-1(IV) chain Protein

A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(IV) chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8716-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001290039.1
Collagen alpha-1(IV) chain Protein
Research Use Only
COL4A1
Recombinant Protein
E. coli
GCAGSGCGKC DCHGVKGQKG ERGLPGLQGV IGFPGMQGPE GPQGPPGQKG DTGEPGLPGT KGTRGPPGAS GYPGNPGLPG IPGQDGPPGP PGIPGCNGTK GERGPLGPPG LPGFAGNPGP PGLPGMKGDP GEILGHVP
Greater than 90% as determined by SDS-PAGE.
1282
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers