missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(IV) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(IV) chain, was expressed in the hosts and tags indicated.
160.00 EUR - 1855.00 EUR
Specifications
Accession Number | NP_001290039.1 |
---|---|
Gene ID (Entrez) | 1282 |
Name | Collagen alpha-1(IV) chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026101
|
enQuireBio™
QP8716-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076101
|
enQuireBio™
QP8716-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036101
|
enQuireBio™
QP8716-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056101
|
enQuireBio™
QP8716-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066101
|
enQuireBio™
QP8716-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046101
|
enQuireBio™
QP8716-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001290039.1 | |
Collagen alpha-1(IV) chain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1282 | |
Research Use Only | |
COL4A1 | |
Recombinant Protein | |
E. coli | |
GCAGSGCGKC DCHGVKGQKG ERGLPGLQGV IGFPGMQGPE GPQGPPGQKG DTGEPGLPGT KGTRGPPGAS GYPGNPGLPG IPGQDGPPGP PGIPGCNGTK GERGPLGPPG LPGFAGNPGP PGLPGMKGDP GEILGHVP | |
Greater than 90% as determined by SDS-PAGE. |