missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Centrosomal protein of 126 kDa Protein

A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 126 kDa, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8097-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052651

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_065853.3
Centrosomal protein of 126 kDa Protein
Research Use Only
KIAA1377
Recombinant Protein
E. coli
HKKMKYNIHE RNGVRFLKSI LKKESKYEHG YLKALIINQS FKFGNQKAAA IRDSIELTKE KGAEIPKTIK KLRWFDETSN IENNAENSHS LKNKTGTTQQ HSQQFHIQSG AG
Greater than 90% as determined by SDS-PAGE.
57562
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers