missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrosomal protein of 126 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 126 kDa, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_065853.3 |
---|---|
Gene ID (Entrez) | 57562 |
Name | Centrosomal protein of 126 kDa Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042651
|
enQuireBio™
QP8097-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062651
|
enQuireBio™
QP8097-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052651
|
enQuireBio™
QP8097-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072651
|
enQuireBio™
QP8097-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082651
|
enQuireBio™
QP8097-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092651
|
enQuireBio™
QP8097-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_065853.3 | |
Centrosomal protein of 126 kDa Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
57562 | |
Research Use Only | |
KIAA1377 | |
Recombinant Protein | |
E. coli | |
HKKMKYNIHE RNGVRFLKSI LKKESKYEHG YLKALIINQS FKFGNQKAAA IRDSIELTKE KGAEIPKTIK KLRWFDETSN IENNAENSHS LKNKTGTTQQ HSQQFHIQSG AG | |
Greater than 90% as determined by SDS-PAGE. |