missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD300e/LMIR6 Fc Chimera Protein

Product Code. 18738316
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738316 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18738316 Supplier R&D Systems™ Supplier No. 10479CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells.

The ED50 for this effect 0.6-6.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q496F6.2
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 342510
Molecular Weight (g/mol) Observed: 45-56 kDa, under reducing conditions; Theroretical: 44 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human CD300e/LMIR6 protein Human CD300e/LMIR6 (Leu18-Leu169) Accession # Q496F6.2(N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Protein CD300e/LMIR6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.