Learn More
enQuireBio™ Recombinant Human Cathepsin F Protein
Product Code. 16016701
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16016701
Brand: enQuireBio™ QP8829ec100ug
A DNA sequence encoding the Homo sapiens (Human) Cathepsin F, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_003784.2
Gene ID (Entrez) 8722
Name Cathepsin F Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CTSF
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence PEWDWRSKGA VTKVKDQGMC GSCWAFSVTG NVEGQWFLNQ GTLLSLSEQE LLDCDKMDKA CMGGLPSNAY SAIKNLGGLE TEDDYSYQGH MQSCNFSAEK AKVYINDSVE LSQNEQKLAA WLAKRGPISV AINAFGMQFY RHGISRPLRP LCSPWLIDHA VLLVGYGNRS DVPFWAIKNS WGTDWGEKGY YYLHRGSGAC GVNTMASSAV VD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.