missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CANT1 Protein
A DNA sequence encoding the Homo sapiens (Human) CANT1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8754-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001153244.1 | |
CANT1 Protein | |
Research Use Only | |
CANT1 | |
Recombinant Protein | |
E. coli | |
GQAPANWYND TYPLSPPQRT PAGIRYRIAV IADLDTESRA QEENTWFSYL KKGYLTLSDS GDKVAVEWDK DHGVLESHLA EKGRGMELSD LIVFNGKLYS VDDRTGVVYQ IEGSKAVPWV ILSDGDGTVE KGFKAEWLAV KDERLYVGGL GKEWTTTTGD VVNENPEWVK VVGYKGSVDH ENWVSNYNAL RAAAGIQPPG YLIHESACWS DTLQRWFFLP RRASQERYSE KDDERKGANL LLSASPDFGD IAVSHVGAVV PTHGFSSFKF IPNTDDQIIV ALKSEEDSGR VASYIMAFTL DGRFLLPETK IGSVKYEG | |
Greater than 90% as determined by SDS-PAGE. |
124583 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |