missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CANT1 Protein
A DNA sequence encoding the Homo sapiens (Human) CANT1, was expressed in the hosts and tags indicated.
160.00 EUR - 1855.00 EUR
Specifications
Accession Number | NP_001153244.1 |
---|---|
Gene ID (Entrez) | 124583 |
Name | CANT1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16076301
|
enQuireBio™
QP8754-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016311
|
enQuireBio™
QP8754-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066301
|
enQuireBio™
QP8754-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096301
|
enQuireBio™
QP8754-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006311
|
enQuireBio™
QP8754-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086301
|
enQuireBio™
QP8754-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001153244.1 | |
CANT1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
124583 | |
Research Use Only | |
CANT1 | |
Recombinant Protein | |
E. coli | |
GQAPANWYND TYPLSPPQRT PAGIRYRIAV IADLDTESRA QEENTWFSYL KKGYLTLSDS GDKVAVEWDK DHGVLESHLA EKGRGMELSD LIVFNGKLYS VDDRTGVVYQ IEGSKAVPWV ILSDGDGTVE KGFKAEWLAV KDERLYVGGL GKEWTTTTGD VVNENPEWVK VVGYKGSVDH ENWVSNYNAL RAAAGIQPPG YLIHESACWS DTLQRWFFLP RRASQERYSE KDDERKGANL LLSASPDFGD IAVSHVGAVV PTHGFSSFKF IPNTDDQIIV ALKSEEDSGR VASYIMAFTL DGRFLLPETK IGSVKYEG | |
Greater than 90% as determined by SDS-PAGE. |