missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-1 adrenergic receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Beta-1 adrenergic receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8958-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007311

  • 416.00 EUR / 50µg


Estimated Shipment
07-08-2020
Add to basket

Specifications

Specifications

NP_000675.1
Beta-1 adrenergic receptor Protein
50 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
153
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADRB1
Recombinant Protein
Yeast
CRSPDFRKAF QRLLCCARRA ARRRHATHGD RPRASGCLAR PGPPPSPGAA SDDDDDDVVG ATPPARLLEP WAGCNGGAAA DSDSSLDEPC RPGFASESKV
Videos
Documents